Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NK2 homeobox 4 | NKX2-4 |
NKX2.4, NKX2D |
-101.9kb | Sense | Chr. 20 - 21395367 - 21397409 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929625 | LOC101929625 | 67.1kb | Anti-sense | Chr. 20 - 21570024 - 21615722 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map