Assay Details
| Gene Symbol: NBAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126995.1 | - | 1 | 3249 | 97 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB305307.1 | - | 1 | Chr. 2 - 15166917 - 15167443 [+] on Build GRCh38 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC653602 | LOC653602 | -516.1kb | Anti-sense | Chr. 2 - 14635091 - 14650819 on NCBI Build GRCh38 | |
| Downstream | DEAD/H-box helicase 1 | DDX1 |
DBP-RB, UKVH5d |
421.0kb | Anti-sense | Chr. 2 - 15591621 - 15631111 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NBAS
Genomic Map