Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927987 | LOC101927987 | -92.6kb | Anti-sense | Chr. 2 - 79493716 - 79500844 on NCBI Build GRCh38 | |
| Downstream | microRNA 4264 | MIR4264 | 51.8kb | Anti-sense | Chr. 2 - 79649294 - 79649359 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map