Assay Details
| Gene Symbol: ZSCAN12P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127188.1 | - | 1 | 1738 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 165 | ZNF165 |
CT53, LD65, ZSCAN7 |
-4.6kb | Sense | Chr. 6 - 28080704 - 28104244 on NCBI Build GRCh38 |
| Downstream | ZSCAN16 antisense RNA 1 | ZSCAN16-AS1 | 24.9kb | Anti-sense | Chr. 6 - 28121795 - 28137316 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZSCAN12P1
Genomic Map