Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NTM antisense RNA 1 | NTM-AS1 | C11orf39 | -74.3kb | Anti-sense | Chr. 11 - 131662129 - 131663556 on NCBI Build GRCh38 |
| Downstream | NTM intronic transcript | NTM-IT | NTM-IT3 | 542.1kb | Sense | Chr. 11 - 132284302 - 132288459 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map