Assay Details
| Gene Symbol: SRGAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104189.1 | 2 - 3 | 328 | 57 | ||
| GenBank mRNA | AK127439.1 | - | 2 - 3 | 328 | 57 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 72 member B | FAM72B | p17 | -175.9kb | Sense | Chr. 1 - 121167646 - 121186896 on NCBI Build GRCh38 |
| Downstream | embigin pseudogene 1 | EMBP1 | 121.4kb | Anti-sense | Chr. 1 - 121519112 - 121571888 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008208.hg.1 | ENST00000581653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008208.hg.1 | ENST00000581666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008208.hg.1 | ENST00000581666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008208.hg.1 | ENST00000581666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015526.hg.1 | ENST00000456469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015526.hg.1 | ENST00000456469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015526.hg.1 | ENST00000456469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017098.hg.1 | ENST00000450872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017098.hg.1 | ENST00000450872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007595.hg.1 | ENST00000569859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215845.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215845.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215845.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215845.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215845.hg.1 | TC0100015511.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011530.hg.1 | ENST00000564795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009716.hg.1 | ENST00000441760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015526.hg.1 | ENST00000456469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015526.hg.1 | ENST00000456469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015526.hg.1 | ENST00000456469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008208.hg.1 | ENST00000581653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008208.hg.1 | ENST00000581666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008208.hg.1 | ENST00000581666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008208.hg.1 | ENST00000581666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215847.hg.1 | TC0100015511.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017098.hg.1 | ENST00000450872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017098.hg.1 | ENST00000450872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000563904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000564145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011528.hg.1 | ENST00000570151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015511.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | ENST00000429055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215848.hg.1 | TC0100015511.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100215849.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215849.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215849.hg.1 | ENST00000437515.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215849.hg.1 | NR_104189.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215849.hg.1 | TC0100015511.hg.1 | Clariom D TC |

SRGAP2-AS1
Genomic Map