Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
unc-80 homolog, NALCN activator |
UNC80 |
C2orf21, UNC-80 |
Sense | Chr. 2 - 209771925 - 209999300 on NCBI Build GRCh38 | |
| Downstream | ribulose-5-phosphate-3-epimerase | RPE | RPE2-1 | 14.8kb | Sense | Chr. 2 - 210002565 - 210029156 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map