Assay Details
| Gene Symbol: LOC100130587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110634.1 | 4 - 5 | 2950 | 87 | ||
| GenBank mRNA | AK127527.1 | - | 4 - 5 | 2950 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723788 | LOC102723788 | Anti-sense | Chr. 20 - 63370753 - 63378135 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC100130152 | LOC100130152 | 21.7kb | Anti-sense | Chr. 20 - 63392887 - 63398343 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3382914 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3303016 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3382919 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20777 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30110 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33001 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3303016 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3382914 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3382919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156278.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156278.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156278.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156278.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156278.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156275.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156275.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156275.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156275.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156275.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156281.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156276.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156276.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156276.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156276.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156276.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156279.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156279.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156279.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156279.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156279.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156280.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156284.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156284.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156284.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156284.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156284.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008061.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | ENST00000428531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156285.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156274.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156274.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156274.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156274.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156274.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156277.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156277.hg.1 | NR_110634.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000156277.hg.1 | TC2000008061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000156277.hg.1 | ENST00000370257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000156277.hg.1 | NR_110634.1 | Refseq |

LOC100130587
Genomic Map