Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1127 | LINC01127 | -4.0kb | Anti-sense | Chr. 2 - 101983465 - 101985545 on NCBI Build GRCh38 | |
| Downstream | interleukin 1 receptor type 2 | IL1R2 |
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB |
158.0 | Anti-sense | Chr. 2 - 101991805 - 102028544 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map