Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell death-inducing DFFA-like effector a | CIDEA | CIDE-A | -1.8kb | Sense | Chr. 18 - 12254319 - 12277595 on NCBI Build GRCh38 |
| Downstream | tubulin beta 6 class V | TUBB6 |
HsT1601, TUBB-5 |
24.9kb | Sense | Chr. 18 - 12307669 - 12329826 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map