Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane and tetratricopeptide repeat containing 3 | TMTC3 | SMILE | -302.0kb | Anti-sense | Chr. 12 - 88142296 - 88199887 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC728084 | LOC728084 | 505.4kb | Sense | Chr. 12 - 89011126 - 89019692 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map