Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized FLJ36000 | FLJ36000 | -21.6kb | Sense | Chr. 17 - 22406019 - 22413741 on NCBI Build GRCh38 | |
| Downstream | MT-RNR2-like 1 | MTRNR2L1 | HN1 | 84.1kb | Sense | Chr. 17 - 22523111 - 22524665 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map