Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369443 | LOC105369443 | -1.6mb | Anti-sense | Chr. 11 - 96508425 - 96514750 on NCBI Build GRCh38 | |
| Downstream | contactin 5 | CNTN5 |
HNB-2s, NB-2 |
845.6kb | Anti-sense | Chr. 11 - 99020904 - 100358885 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map