Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 2 | ZBTB2 | ZNF437 | Sense | Chr. 6 - 151364115 - 151391705 on NCBI Build GRCh38 | |
| Downstream | required for meiotic nuclear division 1 homolog | RMND1 |
C6orf96, COXPD11, RMD1, bA351K16, bA351K16.3 |
32.9kb | Sense | Chr. 6 - 151404548 - 151452181 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map