Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dihydrouridine synthase 2 | DUS2 |
DUS2L, SMM1, URLC8 |
-60.6kb | Sense | Chr. 16 - 68022944 - 68079323 on NCBI Build GRCh38 |
| Downstream | epithelial splicing regulatory protein 2 | ESRP2 | RBM35B | 85.3kb | Anti-sense | Chr. 16 - 68228547 - 68236233 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map