Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box P1 | FOXP1 |
12CC4, HSPC215, MFH, QRF1, hFKH1B |
-954.0 | Anti-sense | Chr. 3 - 70954714 - 71583989 on NCBI Build GRCh38 |
| Downstream | eukaryotic translation initiation factor 4E family member 3 | EIF4E3 |
eIF-4E3, eIF4E-3 |
91.9kb | Anti-sense | Chr. 3 - 71679289 - 71754831 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map