Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 189 | ZNF189 | -8.4kb | Sense | Chr. 9 - 101398830 - 101410660 on NCBI Build GRCh38 | |
| Downstream | TMEM246 antisense RNA 1 | TMEM246-AS1 | 45.6kb | Sense | Chr. 9 - 101468439 - 101481506 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map