Assay Details
| Gene Symbol: LOC101928780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128544.1 | - | 2 - 3 | 3195 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TCF3 fusion partner | TFPT |
FB1, INO80F, amida |
-6.2kb | Sense | Chr. 19 - 54107013 - 54115675 on NCBI Build GRCh38 |
| Downstream | CCR4-NOT transcription complex subunit 3 | CNOT3 |
LENG2, NOT3, NOT3H |
15.3kb | Anti-sense | Chr. 19 - 54137689 - 54155708 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928780
Genomic Map