Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PITPNA antisense RNA 1 | PITPNA-AS1 | -32.4kb | Anti-sense | Chr. 17 - 1516919 - 1518096 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 43 member 2 | SLC43A2 | LAT4 | 18.1kb | Sense | Chr. 17 - 1569254 - 1630014 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map