Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | otoraplin | OTOR |
FDP, MIAL1 |
-604.2kb | Sense | Chr. 20 - 16748353 - 16752164 on NCBI Build GRCh38 |
| Downstream | beaded filament structural protein 1 | BFSP1 |
CP115, CP94, CTRCT33, LIFL-H |
136.0kb | Anti-sense | Chr. 20 - 17493905 - 17569220 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map