Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mediator of cell motility 1 | MEMO1 |
C2orf4, CGI-27, MEMO, NS5ATP7 |
Sense | Chr. 2 - 31867188 - 32011052 on NCBI Build GRCh38 | |
| Downstream |
dpy-30, histone methyltransferase complex regulatory subunit |
DPY30 |
Cps25, HDPY-30, Saf19 |
145.1kb | Sense | Chr. 2 - 32011649 - 32039840 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map