Assay Details
| Gene Symbol: LOC101929457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135692.1 | 1 - 2 | 746 | 64 | ||
| GenBank mRNA | BC062542.1 | - | 1 - 2 | 746 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | high mobility group 20A | HMG20A |
HMGX1, HMGXB1 |
-40.0kb | Sense | Chr. 15 - 77420651 - 77519899 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat and Ig domain containing 1 | LINGO1 |
LERN1, LRRN6A, UNQ201 |
78.9kb | Anti-sense | Chr. 15 - 77613024 - 77820900 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929457
Genomic Map