Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 37B pseudogene 1 | LRRC37BP1 | LRRC37B2 | -74.2kb | Sense | Chr. 17 - 30631755 - 30637477 on NCBI Build GRCh38 |
| Downstream | cytokine receptor like factor 3 | CRLF3 |
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2 |
69.2kb | Anti-sense | Chr. 17 - 30782684 - 30824760 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map