Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kelch repeat and BTB domain containing 2 | KBTBD2 | BKLHD1 | -21.6kb | Sense | Chr. 7 - 32868166 - 32892144 on NCBI Build GRCh38 |
| Downstream | retinitis pigmentosa 9 pseudogene | RP9P | 1.7kb | Sense | Chr. 7 - 32916815 - 32943170 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map