Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | follistatin | FST | FS | -182.9kb | Sense | Chr. 5 - 53480338 - 53487134 on NCBI Build GRCh38 |
| Downstream | ADP ribosylation factor like GTPase 15 | ARL15 | ARFRP2 | 213.5kb | Anti-sense | Chr. 5 - 53883942 - 54310590 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map