Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 689 | ZNF689 | TIPUH1 | -43.3kb | Sense | Chr. 16 - 30602558 - 30610775 on NCBI Build GRCh38 |
| Downstream | fibrosin | FBRS |
FBS, FBS1 |
4.1kb | Anti-sense | Chr. 16 - 30658142 - 30670810 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map