Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bestrophin 1 | BEST1 |
ARB, BEST, BMD, RP50, TU15B, VMD2 |
Anti-sense | Chr. 11 - 61946722 - 61964465 on NCBI Build GRCh38 | |
| Downstream | ferritin heavy chain 1 | FTH1 |
FHC, FTH, FTHL6, HFE5, PIG15, PLIF |
9.7kb | Sense | Chr. 11 - 61964285 - 61967660 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map