| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000570901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000573447.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
ENST00000577163.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011471.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
NR_110900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
TC1600011471.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000571449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000571963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000575089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
ENST00000576590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600192401.hg.1 |
NR_110900.1 |
Refseq |