Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal L1 domain containing 1 | RSL1D1 |
CSIG, L12, PBK1, UTP30 |
-29.5kb | Sense | Chr. 16 - 11834196 - 11851585 on NCBI Build GRCh38 |
| Downstream | nuclear pore complex interacting protein family member A1 pseudogene | LOC729978 | 44.7kb | Sense | Chr. 16 - 11927245 - 11942045 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map