Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translocase of outer mitochondrial membrane 40 | TOMM40 |
C19orf1, D19S1177E, PER-EC1, PEREC1, TOM40 |
-5.7kb | Sense | Chr. 19 - 44891220 - 44903689 on NCBI Build GRCh38 |
| Downstream | apolipoprotein C1 pseudogene 1 | APOC1P1 | 11.8kb | Sense | Chr. 19 - 44926803 - 44931386 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map