Assay Details
| Gene Symbol: RWDD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322335.1 | NP_001309264.1 | 3 | 2194 | 91 | |
| NM_001322336.1 | NP_001309265.1 | 4 | 2505 | 91 | ||
| NM_001322337.1 | NP_001309266.1 | 4 | 2490 | 91 | ||
| NM_001322339.1 | NP_001309268.1 | 4 | 2286 | 91 | ||
| NM_033411.4 | NP_219479.2 | 3 | 2413 | 91 | ||
| XM_017010222.1 | XP_016865711.1 | 3 | 2996 | 91 | ||
| XM_017010223.1 | XP_016865712.1 | 3 | 2771 | 91 | ||
| GenBank mRNA | AJ420581.1 | - | 1 | 552 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoglucomutase 3 | PGM3 |
AGM1, IMD23, PAGM, PGM 3 |
-3.1kb | Anti-sense | Chr. 6 - 83150728 - 83193936 on NCBI Build GRCh38 |
| Downstream | malic enzyme 1 | ME1 |
HUMNDME, MES |
11.5kb | Anti-sense | Chr. 6 - 83210389 - 83431219 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RWDD2A
Genomic Map