Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BIRC6 antisense RNA 2 | BIRC6-AS2 |
linc-birc6, megamind |
Anti-sense | Chr. 2 - 32557520 - 32574818 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 27 | TTC27 | 61.6kb | Sense | Chr. 2 - 32628020 - 32821051 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map