Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane associated ring-CH-type finger 2 | MARCH2 |
HSPC240, MARCH-II, RNF172 |
-31.6kb | Anti-sense | Chr. 19 - 8413272 - 8439021 on NCBI Build GRCh38 |
| Downstream | PML-RARA regulated adaptor molecule 1 | PRAM1 |
PML-RAR, PRAM-1 |
17.9kb | Sense | Chr. 19 - 8490055 - 8502654 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map