Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 173 | TMEM173 |
ERIS, MITA, MPYS, NET23, SAVI, STING, hMITA, hSTING |
-81.2kb | Anti-sense | Chr. 5 - 139475528 - 139482790 on NCBI Build GRCh38 |
| Downstream | CXXC finger protein 5 | CXXC5 |
CF5, HSPC195, RINF, WID |
81.8kb | Sense | Chr. 5 - 139647299 - 139683885 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map