Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TVP23C-CDRT4 readthrough | TVP23C-CDRT4 | FAM18B2-CDRT4 | Sense | Chr. 17 - 15436015 - 15563631 on NCBI Build GRCh38 | |
| Downstream | trans-golgi network vesicle protein 23 homolog C (S. cerevisiae) | TVP23C | FAM18B2 | 24.1kb | Sense | Chr. 17 - 15502264 - 15563631 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map