Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7705 | MIR7705 | hsa-mir-7705 | -13.0kb | Sense | Chr. 8 - 100702968 - 100703024 on NCBI Build GRCh38 |
| Downstream | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta | YWHAZ |
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD |
200.9kb | Sense | Chr. 8 - 100918576 - 100954068 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map