Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NudC domain containing 1 | NUDCD1 |
CML66, OVA66 |
Anti-sense | Chr. 8 - 109240919 - 109334205 on NCBI Build GRCh38 | |
| Downstream |
ENY2, transcription and export complex 2 subunit |
ENY2 |
DC6, Sus1, e(y)2 |
17.8kb | Sense | Chr. 8 - 109334323 - 109345960 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map