Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4319 | MIR4319 | -15.9kb | Anti-sense | Chr. 18 - 44970082 - 44970166 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 14 member 2 | SLC14A2 |
HUT2, UT-A2, UT2, UTA, UTR, hUT-A6 |
223.8kb | Sense | Chr. 18 - 45196357 - 45683686 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map