Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929280 | LOC101929280 | -4.8kb | Sense | Chr. 16 - 594997 - 597964 on NCBI Build GRCh38 | |
| Downstream |
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1 |
WFIKKN1 |
C16orf12, RJD2, WFDC20A, WFIKKN |
23.5kb | Sense | Chr. 16 - 631012 - 634116 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map