Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 173 | GPR173 | SREB3 | Sense | Chr. X - 53049324 - 53080615 on NCBI Build GRCh38 | |
| Downstream | TSPY like 2 | TSPYL2 |
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX |
24.8kb | Sense | Chr. X - 53082327 - 53088546 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map