Assay Details
| Gene Symbol: LOC105371022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093043.1 | - | 2 | 2055 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADAM metallopeptidase with thrombospondin type 1 motif 17 | ADAMTS17 | Anti-sense | Chr. 15 - 99971438 - 100342924 on NCBI Build GRCh38 | ||
| Downstream | spermatogenesis associated 41 (non-protein coding) | SPATA41 | HSD47 | 363.7kb | Anti-sense | Chr. 15 - 100344457 - 100350718 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105371022
Genomic Map