Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC646522 | LOC646522 | Sense | Chr. 11 - 133783671 - 133810376 on NCBI Build GRCh38 | ||
| Downstream | spermatogenesis associated 19 | SPATA19 |
CT132, SPAS1, spergen1 |
35.6kb | Sense | Chr. 11 - 133835930 - 133845538 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map