Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 71 member A | FAM71A | Anti-sense | Chr. 1 - 212624447 - 212626778 on NCBI Build GRCh38 | ||
| Downstream | basic leucine zipper ATF-like transcription factor 3 | BATF3 |
JDP1, JUNDM1, SNFT |
61.4kb | Sense | Chr. 1 - 212686417 - 212699985 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map