Assay Details
| Gene Symbol: IFT52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097603.1 | - | 2 | 1253 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
SGK2, serine/threonine kinase 2 |
SGK2 |
H-SGK2, dJ138B7.2 |
-60.4kb | Sense | Chr. 20 - 43558995 - 43585633 on NCBI Build GRCh38 |
| Downstream | MYB proto-oncogene like 2 | MYBL2 |
B-MYB, BMYB |
19.8kb | Sense | Chr. 20 - 43667019 - 43716496 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

IFT52
Genomic Map