Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SOCS2 antisense RNA 1 | SOCS2-AS1 | Sense | Chr. 12 - 93565628 - 93571398 on NCBI Build GRCh38 | ||
| Downstream | CASP2 and RIPK1 domain containing adaptor with death domain | CRADD |
MRT34, RAIDD |
106.4kb | Anti-sense | Chr. 12 - 93677342 - 93894840 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map