Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cervical cancer-associated transcript 37 | CRAT37 | -666.7kb | Sense | Chr. 15 - 91463339 - 91494850 on NCBI Build GRCh38 | |
| Downstream | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2 | ST8SIA2 |
HsT19690, SIAT8B, ST8SIA-II, STX |
230.4kb | Sense | Chr. 15 - 92393910 - 92468728 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map