Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterile alpha motif domain containing 12 | SAMD12 | Sense | Chr. 8 - 118131828 - 118621995 on NCBI Build GRCh38 | ||
| Downstream | SAMD12 antisense RNA 1 | SAMD12-AS1 |
C8orf26, NCRNA00252 |
360.3kb | Anti-sense | Chr. 8 - 118621001 - 118726067 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map