Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PSMD6 antisense RNA 2 | PSMD6-AS2 | Sense | Chr. 3 - 64004022 - 64012243 on NCBI Build GRCh38 | ||
| Downstream | PRICKLE2 antisense RNA 1 | PRICKLE2-AS1 | 55.9kb | Sense | Chr. 3 - 64067964 - 64103131 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map