Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L19 | MRPL19 |
L19mt, MRP-L15, MRP-L19, MRPL15, RLX1, RPML15 |
-9.2kb | Sense | Chr. 2 - 75646783 - 75662208 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat transmembrane neuronal 4 | LRRTM4 | 1.1mb | Anti-sense | Chr. 2 - 76747723 - 77522433 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map