Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
syncoilin, intermediate filament protein |
SYNC |
SYNC1, SYNCOILIN |
Anti-sense | Chr. 1 - 32679906 - 32702760 on NCBI Build GRCh38 | |
| Downstream | KIAA1522 | KIAA1522 | 52.9kb | Sense | Chr. 1 - 32741839 - 32774970 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map